Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059322.1 | XP_017914811.1 | 4 - 5 | 659 | 97 | |
XM_018059323.1 | XP_017914812.1 | 5 - 6 | 411 | 97 | ||
GenBank mRNA | JT724187.1 | - | 6 - 7 | 2332 | 97 |
Gene Symbol: LOC108637769 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060239.1 | XP_017915728.1 | 2 | 614 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |