Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064074.1 | XP_017919563.1 | 3 - 4 | 1092 | 82 | |
XM_018064075.1 | XP_017919564.1 | 3 - 4 | 581 | 82 | ||
GenBank mRNA | JO645222.2 | - | 1 - 2 | 1441 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |