Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058336.1 | XP_017913825.1 | 6 - 7 | 1837 | 69 | |
XM_018058337.1 | XP_017913826.1 | 5 - 6 | 1780 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |