Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675132.3 | XP_005675189.1 | 3 - 4 | 561 | 81 | |
XM_018046685.1 | XP_017902174.1 | 4 - 5 | 660 | 81 | ||
GenBank mRNA | KA331151.1 | - | 3 - 4 | 525 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |