Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285713.1 | NP_001272642.1 | 1 - 2 | 159 | 99 | |
GenBank mRNA | AF000167.1 | - | 1 - 2 | 159 | 99 | |
JO333451.2 | - | 5 - 6 | 850 | 99 | ||
JO439303.2 | - | 5 - 6 | 936 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |