Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285551.1 | NP_001272480.1 | 2 - 3 | 236 | 78 | |
GenBank mRNA | HQ285239.1 | - | 2 - 3 | 236 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |