Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285585.1 | NP_001272514.1 | 2 - 3 | 87 | 58 | |
XM_013964673.2 | XP_013820127.2 | 2 - 3 | 78 | 58 | ||
XM_013964674.2 | XP_013820128.2 | 2 - 3 | 137 | 58 | ||
XM_013964675.2 | XP_013820129.2 | 2 - 3 | 137 | 58 | ||
XM_013964677.2 | XP_013820131.2 | 2 - 3 | 137 | 58 | ||
GenBank mRNA | AJ249995.1 | - | 1 - 2 | 57 | 58 | |
AJ289715.1 | - | 1 - 2 | 48 | 58 | ||
X65160.1 | - | 2 - 3 | 87 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |