Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289948.1 | NP_001276877.1 | 3 - 4 | 366 | 90 | |
GenBank mRNA | JO352263.2 | - | 2 - 3 | 563 | 90 | |
JO388690.2 | - | 2 - 3 | 577 | 90 | ||
JO446296.2 | - | 2 - 3 | 276 | 90 | ||
JO526704.3 | - | 1 - 2 | 121 | 90 | ||
JO548702.2 | - | 2 - 3 | 509 | 90 | ||
JO565982.2 | - | 2 - 3 | 306 | 90 | ||
JO575245.2 | - | 2 - 3 | 306 | 90 | ||
JO586623.2 | - | 2 - 3 | 540 | 90 | ||
JT738521.1 | - | 2 - 3 | 306 | 90 | ||
KA327599.1 | - | 2 - 3 | 592 | 90 | ||
KC537060.1 | - | 3 - 4 | 366 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |