Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314257.1 | NP_001301186.1 | 1 - 2 | 80 | 88 | |
XM_005681307.3 | XP_005681364.1 | 2 - 3 | 220 | 88 | ||
XM_005681308.3 | XP_005681365.1 | 2 - 3 | 291 | 88 | ||
GenBank mRNA | KA337802.1 | - | 3 - 4 | 3168 | 88 | |
KF667508.1 | - | 1 - 2 | 80 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |