Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314319.1 | NP_001301248.1 | 7 - 8 | 1322 | 117 | |
XM_018042148.1 | XP_017897637.1 | 4 - 5 | 2730 | 117 | ||
GenBank mRNA | KP330489.1 | - | 7 - 8 | 1322 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |