Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285557.1 | NP_001272486.1 | 7 - 8 | 1161 | 74 | |
XM_013970074.2 | XP_013825528.2 | 8 - 9 | 1299 | 74 | ||
XM_013970075.2 | XP_013825529.2 | 8 - 9 | 1180 | 74 | ||
XM_018059920.1 | XP_017915409.1 | 8 - 9 | 1289 | 74 | ||
GenBank mRNA | HQ326238.1 | - | 7 - 8 | 1164 | 74 | |
JO419527.2 | - | 9 - 10 | 1467 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |