Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285610.1 | NP_001272539.1 | 6 - 7 | 516 | 92 | |
XM_018056279.1 | XP_017911768.1 | 7 - 8 | 923 | 92 | ||
XM_018056280.1 | XP_017911769.1 | 7 - 8 | 911 | 92 | ||
XM_018056281.1 | XP_017911770.1 | 7 - 8 | 926 | 92 | ||
GenBank mRNA | EF105409.1 | - | 6 - 7 | 516 | 92 | |
JT699976.1 | - | 4 - 5 | 492 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |