Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285745.1 | NP_001272674.1 | 6 - 7 | 583 | 97 | |
GenBank mRNA | GU985269.1 | - | 6 - 7 | 583 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |