Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314233.1 | NP_001301162.1 | 2 - 3 | 412 | 85 | |
XM_005689565.3 | XP_005689622.1 | 3 - 4 | 545 | 85 | ||
GenBank mRNA | JO579495.1 | - | 6 - 7 | 1835 | 85 | |
JQ990219.1 | - | 2 - 3 | 412 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |