Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314293.1 | NP_001301222.1 | 5 - 6 | 517 | 71 | |
GenBank mRNA | KM369171.1 | - | 5 - 6 | 517 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |