Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314293.1 | NP_001301222.1 | 8 - 9 | 1005 | 117 | |
GenBank mRNA | KM369171.1 | - | 8 - 9 | 1005 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |