Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285595.1 | NP_001272524.1 | 2 - 3 | 148 | 83 | |
XM_018049969.1 | XP_017905458.1 | 2 - 3 | 178 | 83 | ||
GenBank mRNA | HQ846826.1 | - | 2 - 3 | 148 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |