Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001303466.1 | NP_001290395.1 | 3 - 4 | 356 | 65 | |
XM_013964233.2 | XP_013819687.1 | 4 - 5 | 454 | 65 | ||
GenBank mRNA | JO435970.2 | - | 3 - 4 | 565 | 65 | |
KF483978.1 | - | 3 - 4 | 356 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |