Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285618.1 | NP_001272547.1 | 6 - 7 | 958 | 74 | |
XM_013974033.2 | XP_013829487.1 | 7 - 8 | 1077 | 74 | ||
XM_013974034.2 | XP_013829488.1 | 7 - 8 | 1087 | 74 | ||
XM_013974035.2 | XP_013829489.1 | 6 - 7 | 928 | 74 | ||
XM_013974036.2 | XP_013829490.1 | 6 - 7 | 927 | 74 | ||
GenBank mRNA | EF102891.1 | - | 6 - 7 | 958 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |