Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285690.1 | NP_001272619.1 | 3 - 4 | 496 | 93 | |
GenBank mRNA | JO373133.2 | - | 3 - 4 | 508 | 93 | |
U17694.1 | - | 3 - 4 | 496 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |