Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285709.1 | NP_001272638.1 | 1 - 2 | 542 | 88 | |
GenBank mRNA | GQ293360.1 | - | 1 - 2 | 542 | 88 | |
JO419837.2 | - | 6 - 7 | 1126 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |