Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314305.1 | NP_001301234.1 | 7 - 8 | 936 | 75 | |
XM_018058853.1 | XP_017914342.1 | 7 - 8 | 933 | 75 | ||
GenBank mRNA | HM566448.1 | - | 7 - 8 | 937 | 75 | |
KA327728.1 | - | 7 - 8 | 1120 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |