Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285600.2 | NP_001272529.1 | 11 - 12 | 947 | 107 | |
GenBank mRNA | JN942971.1 | - | 11 - 12 | 947 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |