Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285600.2 | NP_001272529.1 | 6 - 7 | 503 | 70 | |
GenBank mRNA | JN942971.1 | - | 6 - 7 | 503 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |