Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285759.1 | NP_001272688.1 | 6 - 7 | 809 | 103 | |
GenBank mRNA | AY389343.1 | - | 6 - 7 | 809 | 103 | |
EF199763.1 | - | 6 - 7 | 772 | 103 | ||
GQ369727.1 | - | 6 - 7 | 760 | 103 | ||
JN377801.1 | - | 5 - 6 | 715 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |