Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314142.1 | NP_001301071.1 | 3 - 4 | 634 | 73 | |
XM_018061650.1 | XP_017917139.1 | 3 - 4 | 1519 | 73 | ||
GenBank mRNA | JQ085593.1 | - | 3 - 4 | 634 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |