Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287558.1 | NP_001274487.1 | 2 - 3 | 244 | 71 | |
GenBank mRNA | HQ606485.1 | - | 2 - 3 | 254 | 71 | |
JO420454.2 | - | 2 - 3 | 219 | 71 | ||
JQ031286.1 | - | 2 - 3 | 244 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |