Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285734.1 | NP_001272663.1 | 3 - 4 | 198 | 99 | |
XM_013963668.2 | XP_013819122.2 | 3 - 4 | 237 | 99 | ||
XM_018047548.1 | XP_017903037.1 | 3 - 4 | 237 | 99 | ||
XM_018047549.1 | XP_017903038.1 | 3 - 4 | 280 | 99 | ||
XM_018047550.1 | XP_017903039.1 | 4 - 5 | 275 | 99 | ||
GenBank mRNA | JN942972.1 | - | 3 - 4 | 198 | 99 | |
JO579731.1 | - | 3 - 4 | 274 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |