Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285734.1 | NP_001272663.1 | 7 - 8 | 469 | 88 | |
XM_013963668.2 | XP_013819122.2 | 7 - 8 | 508 | 88 | ||
GenBank mRNA | JN942972.1 | - | 7 - 8 | 469 | 88 | |
JO579731.1 | - | 7 - 8 | 545 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |