Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285754.1 | NP_001272683.1 | 4 - 5 | 304 | 70 | |
GenBank mRNA | JN107562.1 | - | 4 - 5 | 304 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |