Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696305.3 | XP_005696362.1 | 2 - 3 | 543 | 83 | |
XM_018038733.1 | XP_017894222.1 | 2 - 3 | 607 | 83 | ||
GenBank mRNA | JO429302.2 | - | 3 - 4 | 1466 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |