Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695622.3 | XP_005695679.1 | 4 - 5 | 363 | 65 | |
XM_005695623.3 | XP_005695680.1 | 3 - 4 | 520 | 65 | ||
XM_018038446.1 | XP_017893935.1 | 3 - 4 | 409 | 65 | ||
GenBank mRNA | KA322112.1 | - | 3 - 4 | 319 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |