Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697012.2 | XP_005697069.1 | 1 - 2 | 248 | 91 | |
GenBank mRNA | JP351476.2 | - | 1 - 2 | 126 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |