Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691552.3 | XP_005691609.1 | 2 - 3 | 432 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |