Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052069.1 | XP_017907558.1 | 3 - 4 | 1177 | 80 | |
XM_018052070.1 | XP_017907559.1 | 3 - 4 | 399 | 80 | ||
GenBank mRNA | JO428804.2 | - | 3 - 4 | 1931 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |