Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013964679.2 | XP_013820133.1 | 1 - 2 | 610 | 67 | |
GenBank mRNA | KC236982.1 | - | 1 - 2 | 357 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |