Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319276.1 | NP_001306205.1 | 8 - 9 | 1399 | 60 | |
GenBank mRNA | KR013052.1 | - | 8 - 9 | 1399 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |