Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051920.1 | XP_017907409.1 | 3 - 4 | 366 | 101 | |
XM_018051928.1 | XP_017907417.1 | 3 - 4 | 367 | 101 | ||
XM_018051937.1 | XP_017907426.1 | 3 - 4 | 367 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |