Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049361.1 | XP_017904850.1 | 3 - 4 | 1084 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |