Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696081.3 | XP_005696138.2 | 2 - 3 | 1142 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |