Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050499.1 | XP_017905988.1 | 1 - 2 | 449 | 80 | |
GenBank mRNA | KA318602.1 | - | 1 - 2 | 228 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |