Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060222.1 | XP_017915711.1 | 4 - 5 | 596 | 117 | |
XM_018060223.1 | XP_017915712.1 | 5 - 6 | 783 | 117 | ||
GenBank mRNA | JO433359.2 | - | 5 - 6 | 778 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |