Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046249.1 | XP_017901738.1 | 2 - 3 | 1543 | 84 | |
XM_018046250.1 | XP_017901739.1 | 2 - 3 | 1543 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |