Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690335.3 | XP_005690392.1 | 8 - 9 | 1296 | 90 | |
GenBank mRNA | KA333597.1 | - | 3 - 4 | 422 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |