Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIWIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061075.1 | XP_017916564.1 | 10 - 11 | 1507 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |