Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041077.1 | XP_017896566.1 | 2 - 3 | 520 | 97 | |
GenBank mRNA | KA322484.1 | - | 2 - 3 | 287 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |