Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699761.3 | XP_005699818.1 | 2 - 3 | 367 | 91 | |
GenBank mRNA | JI861816.1 | - | 2 - 3 | 262 | 91 | |
JO439119.2 | - | 2 - 3 | 365 | 91 | ||
JT754263.1 | - | 6 - 7 | 948 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |