Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005709403.3 | XP_005709460.2 | 1 - 2 | 376 | 106 | |
GenBank mRNA | JT679429.1 | - | 3 - 4 | 148 | 106 | |
KA324602.1 | - | 1 - 2 | 321 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |