Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049085.1 | XP_017904574.1 | 3 - 4 | 880 | 79 | |
GenBank mRNA | JO438693.2 | - | 3 - 4 | 801 | 79 | |
JT666219.1 | - | 3 - 4 | 798 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |