Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966271.2 | XP_013821725.2 | 2 - 3 | 2014 | 71 | |
GenBank mRNA | KA328991.1 | - | 2 - 3 | 944 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |